Wall-Mount Eye/Face Washes, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Featuring four GS-Plus “Gentle Spray” heads, these eye/face washes provide a soft, wide stream of water that gently and quickly rinses contaminants away.
- Heavy-duty pipe fittings resist corrosion
The integral, self-adjusting flow controls, located in the head, ensure even distribution of water. Units come complete with a 29.2 cm (11") stainless steel bowl and a 1.3 cm (¹/₂") brass stay-open ball valve.
Certifications: Meet ANSI Z358.1 standard.
Delivery: Supplied with ANSI-compliant safety green emergency eye wash sign.
Customers Who Bought This Also Bought