Deck-Mount Swing-Out Eyewashes, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Featuring the GS-Plus 'Gentle Spray' heads, these eyewashes deliver a soft, wide stream of water to quickly and gently rinse contaminants away.
- Includes self-adjusting flow controls, impact-resistant polypropylene body, and flip-top dust covers
- Auto-flow models are activated when the head assembly is swung from storage to operation position
- An ANSI-compliant eyewash sign is included with all units
Flag-handle models are activated when the head assembly is swung from storage to operation position and the flag handle is pushed. LDPE filter removes scale, deposits, and debris from the water and keeps the water line impurities out of the user’s eyes.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought