Eyesaver® Eyewash Stations, Bar Activation, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
These eyewash stations offer twin aerated sprays for gentle, yet thorough rinsing of the eyes and face.
- Available in deck-mount, wall-mount, and pedestal-mount styles
The units are supplied with a rectangular stainless steel bowl and panic bar activating system.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought