SP Bel-Art Roto-Flo™ Flow Indicators, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These indicators provide immediate visualization of flow in a tubing line.
- Brightly colored paddle wheels spin when gas or liquid flows through a system
- Molded of clear polystyrene
- Smooth connecting barbs for easy installation
- Maximum recommended pressure is 10 psig
Internal flow channel diameter is 5 mm (7/32"), paddle wheel shell is 25 mm (1"), and overall length is 51 mm (2").
Not for use with corrosive chemicals or fluids with temperatures in excess of 64 °C (150 °F).
Spécifications
- Catalog No:
- 47727-238
- 47727-234
- 47727-232
- Connection:
- Barbed
- Barbed
- Barbed
- Min. flow rate:
- 0.5 L/min.
- 0.5 L/min.
- 0.5 L/min.