Stainless Steel Gowning Benches, Metro
Supplier: Metropolitan Wire
Certificates
About this item
Type 304 stainless steel, H.D. Super® Posts and proven Super Erecta construction throughout.
- Heavy-duty, solid top bench
- Sturdy and comfortable
- Type 304 stainless steel
- Adjustable stainless steel foot plates allow floor mounting
- Non-conductive, but can be made conductive with the addition of HD aluminum split sleeves
Heavy-duty, solid top bench provides a sturdy and comfortable growning fixture. Top is Type 304 stainless steel. Adjustable stainless steel foot plates allow floor mounting. Floor mounting required for 9 and 12" wide benches. All benches are 18" (457mm) tall. Unit is non-conductive, but can be made conductive with the addition of HD aluminum split sleeves.
Customers Who Bought This Also Bought