A propos de cet article
Designed for the safe, reliable thawing of fresh frozen plasma and whole blood
- Removable, center-hinged reservoir cover
- 4 °C preset thawing temperature
- Bright temperature and elapsed time displays
- Calibration capability
- Removable, high-efficiency fluid filter
- Reservoir drain
- Over-temperature and flow alarms
Bath includes a built-in timer for elapsed thawing time and a convenient calibration offset function for matching displayed temperature reading to a traceable standard. Built-in self-diagnostics instantly alert users to high temperatures, low liquid flow, and other conditions that may compromise thawing performance or AHF recovery. The reservoir drain valve, reusable high-efficiency liquid filter, and reusable air filter all help minimize maintenance downtime.
Bath provides proper conditions for the recovery of cryoprecipitated antihemophilic factor (AHF). Features a 52.2 L (13.8 gal.) reservoir capable of holding up to 24 units of frozen plasma or whole blood. The reservoir has a large, 33x52.4 cm (13x205/8") opening for easy loading and unloading and comes with a hinged, fully removable cover.
Spécifications
- Catalog No:
- 13272-292
- 13272-290
- Temperature stability:
- ±0.1°C
- ±0.1°C
- Compressor:
- 1/4 hp
- 1/4 hp
- Weight:
- 93 kg (205 lbs.)
- 93 kg (205 lbs.)