Nichipet ECO Glass Tip Pipettors, Nichiryo America
Supplier: Nichiryo America
Certificates
About this item
Designed to provide the accuracy and precision of a micropipettor with the compatibility of glass, the unique Nichipet ECO utilizes reliable and stable glass tips to dispense liquids quickly and safely.
- Pipettors feature molded channels within the handle for improved grip
- Glass tips are autoclavable
- Four different pipettor sizes cover a volume range from 20 µl to 1 ml
Tips are attached to the unit with a double O-ring design, ensuring a secure fit with highly reproducible results. Volume is adjustable through the use of a color-coded dial which locks to prevent volume drift when in use. Pipettors are supplied with a PTFE nozzle and ceramic piston for smooth action.
The ECO is ideal for laboratories dispensing harsh chemicals that require glass for accurate measurements, such as bias-free liquid handling for endocrine disruptor's analysis.
Customers Who Bought This Also Bought