Panel-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Panel-mounted mixing faucets are made of chrome-plated cast brass and provided with a serrated hose end, four-arm handles, and 1 cm (3/8") IPS inlet shanks, locknuts, and washers
Faucets are available with 15.2 cm (6") spread gooseneck, with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought