Curb-Mounted Gooseneck Mixing Faucets, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Curb-mounted mixing faucets are made of chrome-plated cast brass and are provided with a serrated hose end, four-arm handles, and 1 cm (3/8") IPS inlet shanks, locknuts, and washers.
- Faucet is supplied with 20.3 cm (8") spread gooseneck, with or without a vacuum breaker
- Faucet valves are self-contained renewable units with replaceable stainless steel seats
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought