Deck-Mounted Mixing Faucets, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Deck-mounted mixing faucets are made of chrome-plated forged brass and are provided with an aerator, four-arm handles, and 1 cm (3/8") diameter copper tubes on inlets.
Faucets are available with 22.9 cm (9") spread swing spout, with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought