Water Fitting, WaterSaver Faucet
# de catalogue VFT-0306-01
Fournisseur: WaterSaver Faucet
Spécifications
- Description:Water Fitting
- Cat. no.:VFT-0306-01
- Supplier No.:LE2800-131WS
Spécifications
A propos de cet article
Curb- or panel-mounted chrome-plated water fitting consists of a single outlet turret and a straight water stop.
Ordering information: Complete with 9.5mm (3/8") IPS tank nipple, locknut, and washer.
Customers Who Bought This Also Bought