Nalgene® Wide Mouth Centrifuge Bottle with Sealing Cap, Polycarbonate, Conical-Bottom, Thermo Scientific
# de catalogue 21010-608
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:177 ml (6 oz.)
- Ø:61.4 mm
- Length:144.3 mm with cap
- Description:Bottle with Cap
- Cat. no.:21010-608
- Maximum RCF:27500×g
- Supplier No.:3144-0175
Spécifications
A propos de cet article
This transparent centrifuge bottle is manufactured from polycarbonate and is ideal for cell culture and protein biochemistry applications
Note: When autoclaving, place cap on top of container without engaging threads. Bottles must be used with adapter 21020-249. For proper performance, fill bottle to at least 80% of the total capacity.
It is graduated in 25 ml increments. Leakproof and autoclavable.
Ordering information: Supplied with polypropylene sealing cap with retained silicone gasket.
Customers Who Bought This Also Bought