Deck-Mounted Laboratory Ball Valve, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
White epoxy-coated single laboratory ball valve with integral straight serrated hose end.
- Forged brass valve body
- Includes turret base and mounting shank
- Rated for use at pressures up to 75 psi
Certifications: Meets ANSI Z21.15 standard and is AGA/CGA certified.
Ordering information: Please specify service when ordering.
Customers Who Bought This Also Bought