Nalgene® Wide Mouth Centrifuge Bottle with Screw Cap, High-Density Polyethylene, Thermo Scientific
Catalog # 21020-072
Supplier: Thermo Fisher Scientific
Specifications
- Ø:61.2 mm
- Capacity:250 ml (8.5 oz.)
- Description:Centrifuge Bottle with Cap
- Length:131.6 mm
- Cat. no.:21020-072
- Maximum RCF:8000×g
- Supplier No.:3121-0250
Specifications
About this item
Ideal for use in refrigerated centrifuges.
Note: For proper performance, fill bottles to at least 80% of the total capacity.
Wide mouth provides easy access. Withstands temperatures to –100 °C (–148 °F). Manufactured from chemically resistant HDPE. Polypropylene screw closure.
Customers Who Bought This Also Bought