Gas, Air, or Vacuum Fittings, Wall Mounted, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
These wall-mounted faucets are made of chrome-plated brass.
- Maximum Working Pressure: 75 psi
- Index Buttons for Gas, Air, and Vacuum
- Single, Double, or Quadruple Service Available
- Panel Flange or Wye Fitting
- Chrome-Plated Brass Construction
They feature a panel flange or wye fitting and a ball valve for curb or wall mounting.
The faucet includes index buttons for gas, air, and vacuum, and single, double, or quadruple service is available.
Ordering information: This item includes 9.5 mm, (3/8") IPS tank nipples, locknuts, and washers.
Customers Who Bought This Also Bought