NALGENE® White Polypropylene Closures, Thermo Scientific
# de catalogue 16219-120
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:70 mm
- Description:Replacement Closure for 70 mm Jars, White Polypropylene
- Cat. no.:16219-120
- Supplier No.:712154-0700
Spécifications
A propos de cet article
Linerless closures fit NALGENE® Jars 36319-501, 16128-841 series, 16129-306 and -308, 16129-356 and -378
Autoclavable. Cap Size: 70mm.
Customers Who Bought This Also Bought