Electrical Pedestal Boxes, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
The boxes are available in brushed satin finish or wrinkled black epoxy finish. They are furnished with 3-wire grounding receptacle(s) rated for 125 V, 20 A, stainless steel faceplates, grounding screw(s) and 1.9 cm (3/4") mounting shank and locknut.
- Manufactured of cast aluminum
- Machined for both standard and ground fault receptacles
Certifications: UL listed under Standard UL514A.
Customers Who Bought This Also Bought