Boiling Granules, Non-Selenized, Hengar
Fournisseur: Hengar
Certificats
A propos de cet article
These plain granules effectively prevent super-heating of boiling liquids with the accompanying spasmodic, explosive formation of vapors.
- When a few granules are placed in a liquid, boiling proceeds constantly and quietly
- Any gases formed in the liquid are swept out rapidly and completely
- Completely insoluble
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.