Block Digestion Tubes, Tech Glass
Fournisseur: Technical Glass Products
Certificats
A propos de cet article
These borosilicate glass tubes feature a retention lip at the top, rounded bottom, and a graduation mark at the maximum volume level.
Choose from constricted or straight tubes.
Customers Who Bought This Also Bought