Rapid Digestor, 4- and 25-Place, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Units feature individually insulated ceramic heaters, that provide uniform and efficient heat transfer to digestion tubes, RTD platinum sensor, and solid-state stepless temperature controls
Light-monitoring system signals attainment of preset temperature. Accommodates 250ml digestion tubes. Digest samples in 25 to 45 minutes, depending on type or catalyst. Four-place unit (50997-504) is for 115V, 50/60Hz operation. Twenty-five-place unit is for 208/230V, 50/60Hz operation.
Note: Operation in a fume hood is strongly recommended.
Ceramic heater assembly is housed in a chemically etched aluminum casing and resists corrosion.
Ordering information: Digestion tubes and fume removal system are required (sold separately). For fume removal system for 4-place unit, see 50997-606. For fume removal system for 25-place unit, see 51000-507.
Customers Who Bought This Also Bought