Synthware Heat Transfer Block for Round Bottom Flasks, Microscale, Kemtech America
# de catalogue 60004-230
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware heat transfer block for round bottom flasks, microscale
- Supplier No.:H200003
Spécifications
A propos de cet article
Aluminum heat transfer block, used in place of a glass sand bath in microscale experiments.
- Aluminum heat transfer block
- Hold 3 flask sizes
Block holds 5 ml, 10 ml, and 25 ml round bottom flasks, with one hole for a standard thermometer.
Customers Who Bought This Also Bought