UVP Transilluminators, Visi-White™, Analytik Jena
Fournisseur: Analytik Jena US
Certificats
A propos de cet article
View film negatives and positives, autoradiographs, X-ray film, microtiter plates, and stained gels
Glass surface is permanently double safety-sealed into the base for durability. Scratch- and chemical-resistant work surface can be easily cleaned. Housing is finished with an epoxy-powder paint for easy cleaning and maintenance.
Certifications: The 115 V models are ETL and ETL-C approved. The 230V models are CE marked.
Ordering information: Manufacturer’s two-year warranty on all models.
Customers Who Bought This Also Bought