VWR® Modular Heating Blocks, Solid
Fournisseur: VWR International
Certificats
A propos de cet article
Constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
For use as a low-temperature hotplate or for drilling a customized block. Available in single or double block size.
Customers Who Bought This Also Bought