BioRobotix® Pipette Tips for MultiMek™ Workstations, Molecular BioProducts
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Compatible with MultiMek™ style workstations/AP96.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as clear polypropylene only, in 20, 130 and 250 µl sizes. Filter tips utilize the ART barrier for guranteed protection against cross-contamination.
Ordering information: Tips are packaged 96 per tray, 10 trays per pack.
Customers Who Bought This Also Bought