Dry Block Heater, Digital, 240 V Glas-Col®
Fournisseur: Glas-Col
Certificats
A propos de cet article
This small, constant-temperature heater can accommodate a variety of different size blocks
- 240 V
Maintains temperature from slightly above ambient to 125°C (257°F), with ±1°C accuracy. The heater opening is in standard SBS format, which allows heating of high- or low-profile standard well plates. The heater features single-loop, auto-tuning digital control with simple input commands, dual four-digit LED display for temperature in °C or °F, and up and down keys for set point adjustment. The heater also features PID control parameters that set automatically for optimum system performance, an on/off power switch, and fuse protection. The unit exterior is finished in foxtip gray and charcoal.
Ordering information: Heater includes a three-wire cord with molded plug.
Caution: c
Spécifications
- Catalog No:
- 80078-742
- 80078-744
- 80078-746
- 80078-748
- 80078-740
- Dimensions:
- 14.6W x 11.4D x 17.2H cm (53/4 x 41/2 x 63/4")
- Weight:
- 2.3 kg (5 lbs.) Unit