ContactUsLinkComponent
VWR® Urine Sediment Processing Tube
VWR® Urine Sediment Processing Tube
Catalog # 89049-036
Supplier:  VWR International
VWR* Urine Sediment Processing Tube
VWR® Urine Sediment Processing Tube
Catalog # 89049-036
Supplier:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    12 ml
  • Description:
    Urine Sediment Processing Tube
  • Cat. no.:
    89049-036
Specifications
About this item
Disposable polystyrene tubes are designed for use with Sysmex® UF series urine analyzers.
  • Graduated markings
  • Disposable design
  • Polystyrene construction
  • For use with urine analyzers
Graduated to 10 mL, these tubes offer a 12 mL capacity that includes a 400µL collection nipple reservoir. The unique tube design features straight sides that allow for placement in each hole of testing racks. Maximum RCF: 1500g.
Ordering information: Use with cap 82028-226 (ordered separately).
Customers Who Bought This Also Bought