Model 8800 8-Channel Variable Repetitive Syringe Dispenser, Nichiryo America
Fournisseur: Nichiryo America
Certificats
A propos de cet article
The 8800 Syringe Dispenser offers a unique approach to multichannel dispensing.
A choice of replaceable syringes mounted on interchangeable cartridges allow the user to dispense six different volumes of liquid (10, 20, 25, 40, 50, and 100µl) into a microtiter plate. Multiple dispensing can be achieved by repeatedly pressing the end button. The lightweight and slender design allows for easy handling without fatigue, even over extended periods of use. An intuitive adjustment dial provides quick volume identification. Syringes may be quickly and easily interchanged and replaced. Both syringes and cartridges are autoclavable.
Customers Who Bought This Also Bought