Reagent Reservoirs, 12-Channel, Argos Technologies
Supplier: Argos Technologies
Certificates
About this item
These pipetting basins are designed for serial dilutions or pipetting different reagents when utilizing pipettors with up to a 12-channel capacity.
Manufactured from virgin polypropylene, these basins provide excellent chemical resistance and can withstand temperatures from –80 to 121°C (–112 to 250°F). Each of the 12 chambers holds up to 3mL and is V-shaped to facilitate complete sample withdrawal. Each chamber is numbered to ensure accurate sample identification. Basins are available sterile or nonsterile, and are supplied with a fitted lid.
Customers Who Bought This Also Bought