SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Catalog # 74950-313
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Three-In-One Reservoir
- Material:PP
- Cat. no.:74950-313
- Supplier No.:378820000
Specifications
About this item
This versatile reservoir is ideal for use when filling multi-tube blocks or microwell plates.
- Graduated markings
- Suitable for use with all brands of multichannel pipettors
- Autoclavable
It features a graduated single reagent reservoir with graduations at 10, 25, 40, and 50 mL; a reservoir with eight separate 5 mL reagent channels; and a reservoir with 12 separate 5 mL reagent channels. Each compartment of the multichannel reservoirs is numbered for reagent identification.
Customers Who Bought This Also Bought