1536-Well No-Bottom Plates, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Plates are made from virgin polystyrene resin and molded without the bottom of the plate.
- Virgin polystyrene construction
- Designed without a bottom
- Suitable for high-throughput applications
- Durable
HiBase plates are suitable for all standard high-throughput applications.
Customers Who Bought This Also Bought