A propos de cet article
Plates are manufactured from virgin polystyrene resin and are designed to be compatible with microplate handling equipment and robotics.
- Virgin polystyrene construction
- Designed to minimize air bubble formation in wells
- Free of RNase, DNase, and human DNA
- Nonpyrogenic
The square-round well geometry is designed to minimize wicking and reduce air bubble formation, while increasing volume capacity and surface area. Plates are stackable with and without lids. HiBase and LoBase plates are available in LUMITRAC™, FLUOTRAC™, and MICROLON™ formats. LUMITRAC™ format plates are ideal for luminescence assays; FLUOTRAC™ format plates are ideal for fluorescence assays; MICROLON™ format plates are available for consistent performance in binding assays. All plates are RNase-, DNase-, and human DNA-free, and are nonpyrogenic. All plates have footprint conforming to the ANSI/SBS 1-2004 Standard.