1536-Well µClear® Bottom Plates, Greiner Bio-One
Fournisseur: Greiner Bio-One
Certificats
A propos de cet article
Plates are manufactured from virgin polystyrene resin and feature µClear® microthin polystyrene film on the bottom.
- Virgin polystyrene construction
- Free of RNase, DNase, human DNA, and pyrogens
- Designed to reduce air bubble formation in wells
- Stackable with and without lids
Designed to be compatible with microplate handling equipment and robotics
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
The square-round well geometry is designed to minimize wicking and reduce air bubble formation, while increasing volume capacity and surface area. Plates are stackable with and without lids (lids are not included.) HiBase and LoBase plates are available in LUMITRAC™ format for luminescence assays and FLUOTRAC™ format for fluorescence assays.
Customers Who Bought This Also Bought