Cliniplate™ 384-Well Microplates, Thermo Scientific
Catalog # 28298-614
Supplier: Thermo Fisher Scientific
Specifications
- Sterility:Non sterile
- Color:Clear
- Description:Cliniplates™
- Cat. no.:28298-614
- Maximum Well Volume:70 µL
- Supplier No.:95040000
Specifications
About this item
These polystyrene microplates are designed for high throughput screening and are ideal for a wide range of measurement techniques
Clear plates are ideal for photometry applications. White and black plates are designed for assays utilizing fluorescent or luminescent markers. The 384-well format enables higher sample density, lower consumption of reagents, and reduced overall costs. Plates are designed for use with instrumentation adapted for 384-well plates. Cliniplates™ also feature excellent optical performance and rounded wells for improved mixing.
Ordering information: Sterile plates are individually wrapped with lids.
Frequently Bought Together