Spécifications
- Description:96-Well Low-Profile Plate, Clear
- Cat. no.:82050-972
- Maximum Well Volume:8 µL
- Supplier No.:673101
Spécifications
A propos de cet article
Designed to facilitate high throughput microbatch protein crystallography in fully automated systems, these plates enable the investigation of optimal crystal growth conditions for subsequent X-ray diffraction analysis.
- Smooth, flat bottoms for optical clarity
- Low-profile design
- Clear microthin film bottom provides minimized autofluorescence
Wells have conical tops to help center protein droplets for optimized crystallization under oil, and smooth, flat bottoms to assure perfect microscopic visualization of growing crystals. The clear 96-Well Low-Profile Plate is low-profile for minimized storage space. The black 96-Well Plate has a µClear® polystyrene microthin film bottom for clearer imaging or viewing and minimized autofluorescence. The 1536-Well Plate with LBR is made of polyolefin for low birefringence (LBR), and is specially designed for use with polarized light. Plates are supplied without lids.