TipStation Pipette Tip Systems, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
Stackable racks of tips are ideal for use with 8- and 12-channel pipettors.
- Racks, lids, and base are manufactured from recycled plastic
Racks snap together and will not lift during pipetting. Recessed wells keep the tips stable for mounting on pipettors. The stacked design saves bench space and features a wide base to prevent the stack from tipping.
Ordering information: Each rack contains 192 tips in two 8×12 trays. The stack includes five racks of tips, one lid, and one base.
Customers Who Bought This Also Bought