Oxidation Pressure Vessel and Oxidata™ Software, Koehler
# de catalogue 37000-034
Fournisseur: Koehler
Small Business Enterprise
Spécifications
- Description:Oxidation Pressure Vessel
- Cat. no.:37000-034
- Supplier No.:K10500
Spécifications
A propos de cet article
Precision machined stainless steel pressure vessel includes threaded body, lid, stem with filler rod and mounting flange, and needle valve for purging, pressurizing, and exhausting pressure vessel with oxygen
Available Oxidata™ pressure measurement systems (sold separately) run on Windows® XP and offer advanced, real time data acquisition and management for fuel and lubricant ASTM oxidation tests.
Oxidation pressure vessel conforms to specifications: ASTM D525.
Burst disk assembly ruptures at 223psi (1537kPa) to prevent unsafe pressure buildup. Vessel and stem interiors have a high polish to facilitate cleaning and prevent corrosion. Octagonal sections on the vessel and lid permit tight closures with a wrench.
Customers Who Bought This Also Bought