384-Well Deep Well Small Volume™ Microplate, Polypropylene, Greiner Bio-One | Avantor
384-Well Deep Well Small Volume™ Microplate, Polypropylene, Greiner Bio-One
107 µl (21 µl in frustum), 1 - 90 µl
Catalog # 89085-306
Supplier:   Greiner Bio-One
Fournisseur #:  784201
384-Well Deep Well Small Volume™ Microplate, Polypropylene, Greiner Bio-One
Catalog # 89085-306
Supplier:  Greiner Bio-One
384-Well Deep Well Small Volume™ Microplate, Polypropylene, Greiner Bio-One
384-Well Deep Well Small Volume™ Microplate, Polypropylene, Greiner Bio-One
107 µl (21 µl in frustum), 1 - 90 µl
Price
$1,335.76
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 100
Product Details & Documents
The 384 Deep Well Small Volume™ polypropylene microplate combines the advantages of Small Volume™ microplates with the convenience of standard microplate dimensions.
  • Focused liquid samples
  • No loss of valuable compounds
  • Nonsterile
  • Nonpyrogenic
  • Free of RNase, DNase, and human DNA
  • Compatible with automated systems
Plate is especially suited for direct compound transfer and preparation of assay-ready plates, pre-dilutions, heat sealing, and automation
The unique well geometry features a square cross sectional area at the well top that transitions into a truncated cone with a flat base of 1.52mm. The design allows smaller volumes to be aspirated, reduces dead volume, simplifies dilutions, and delivers the most homogenous seal possible for the base of the plate. Color: natural.
Can also be used as a storage plate.
Ordering information: Supplied without lid. For additional information on Small Volume™ plates, see 82051-444 series.
Spécifications
  • Working Volume:
    1 - 90 µl
  • Total Volume:
    107 µl (21 µl in frustum)