Finntip™ Pipette Tips, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
CE Compliant
Certificats
A propos de cet article
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought