SP Bel-Art 12-Channel Mini-Tube Strips, Bel-Art Products, a part of SP
# de catalogue 60917-202
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Mini-Tubes, 400 12-Tube Strips (4,800 Tubes)
- Cat. no.:60917-202
- Supplier No.:378570000
Spécifications
A propos de cet article
These 1 mL volume Mini-Tubes offer a convenient and economical means of performing laboratory procedures using multi-channel pipettors of four, eight, or twelve channels.
- Convenient vessel for laboratory procedures
- Economical
- Easily separated into single tubes
They feature an attachment bridge between tubes, forming a Mini-Tube strip which can be easily separated into single tubes by cutting apart.
Customers Who Bought This Also Bought