Snapwell™ Inserts, Sterile, Corning
Fournisseur: Corning
Certificats
A propos de cet article
A modified Transwell® permeable support containing a 12 mm Ø membrane supported by a detachable ring. Once cells are grown to confluence on the Snapwell™ insert, the ring can be placed in a vertical or horizontal diffusion chamber. Packaged in six well plates.
Ordering information: Snapwell™ inserts are supplied preloaded into 6-well plates.
Spécifications
- Catalog No:
- 29444-282
- 29442-160
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk