ContactUsLinkComponent
1536-Well Acoustic Liquid Handling Storage Plates, Greiner Bio-One
1536-Well Acoustic Liquid Handling Storage Plates, Greiner Bio-One
# de catalogue 89085-692
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
1536 Well Plate
1536-Well Acoustic Liquid Handling Storage Plates, Greiner Bio-One
# de catalogue 89085-692
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Spécifications
  • Color:
    Clear
  • Description:
    Storage Plate with Optimized Geometry for Low Evaporation
  • Working Volume:
    1 - 14 µl
  • Supplier No.:
    792870-906
  • Total Volume:
    16 µl
Spécifications
A propos de cet article
Acoustic liquid dispensing offers the advantage of pipetting small volumes very precisely and with minimal disturbance without the risk of cross-contamination.
  • Solid bottom design
  • Nonsterile
  • Free of DNase, RNase, and human DNA
  • Designed for low evaporation
The process involves the transferral of DMSO-containing solutions in the nanoliter to picoliter range from a starting plate into a target plate by means of ultrasound
All plates are manufactured from cycloolefin and feature solid bottoms and F-bottom well profiles. The LoBase storage plate is ideal for working with minimal sample volumes. The storage plate with optimized geometry for low evaporation is equipped with a groove around the plate edges for a matching cycloolefin plate lid (not included). This prevents evaporation and minimizes edge effects. All plates are nonsterile, nonpyrogenic, and DNase-, RNase-, and human DNA-free.
The plates used have a substantial influence on the accuracy of the amount of liquid transferred. Even wall thicknesses and consistent sealing of the microplate bottoms are essential in maintaining the reproducibility of the sample.
Ordering information: Plates are supplied without lids. Accessory lid is sold separately.
Customers Who Bought This Also Bought