SensoPlate™ 1536-Well Glass-Bottom Plates, Greiner Bio-One
Fournisseur: Greiner Bio-One
Certificats
A propos de cet article
Microplates exhibit a bottom flatness better than 100 µm to offer high-quality performance for all applications where low autofluorescence and exceptional optical clarity are required.
- Ideal for applications requiring low autofluorescence
- Exceptional optical clarity
- Sterile
- Supplied with a lid
Ideal for high-resolution imaging, sensitive fluorescence, and confocal microscopy such as single molecule detection (SMD). Transmission measurements are capable in the wavelength range above 350nm. Plates consist of a 175 µm thick borosilicate glass-bottom plate bonded with a black polystyrene frame. Plates are sterile, comply with SBS recommendations, and include a lid.
Ordering information: For 1536-well SensoPlate™ Plus glass-bottom plates, see 82051-620 series.
Customers Who Bought This Also Bought