SensoPlate™ 24-Well Glass-Bottom Plate, Greiner Bio-One
Catalogue # 82050-898
Fournisseur: Greiner Bio-One
Spécifications
- Color:Black
- Description:SensoPlate™ 24-Well Glass-Bottom Plate
- Cat. no.:82050-898
- Supplier No.:662892
Spécifications
À propos de cet article
Microplate exhibits a bottom flatness better than 100 µm to offer high-quality performance for all applications where low autofluorescence and exceptional optical clarity are required.
- Ideal for applications requiring low autofluorescence
- Exceptional optical clarity
- Sterile
- Chimney-well prevents cross-contamination
Ideal for high-resolution imaging, sensitive fluorescence, and confocal microscopy such as single molecule detection (SMD). Transmission measurements are capable in the wavelength range above 350nm. Plate consists of a 175 µm thick borosilicate glass-bottom plate bonded with a black polystyrene frame and features a chimney-well to help prevent cross-contamination. Plate is sterile, complies with SBS recommendations, and includes a lid.