Corning® Brand 96-Well UV Plates, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Acrylic with UV transparent flat well bottom, non sterile. Ideal for determining protein and/or nucleic acid concentrations.
- UV transparent bottom is moulded directly to an acrylic base for greater strength and maximum leak resistance
- Allows UV absorbance readings with low background, especially at 260 to 280 nm
- Certified free from DNase and RNase
Ordering information: Packed 25 per pack, 50 per case.
Spécifications
- Catalog No:
- 66021-983
- 33501-012
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk