Corning® 96-Well Plates, Polypropylene, Without Lids
Fournisseur: Corning
Certificats
A propos de cet article
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µl plate is packaged 10 plates per bag and 5 bags per case.
Spécifications
- Catalog No:
- 80081-120
- 29444-400
- 29445-150
- 29445-152
- 80081-118
- 29445-112
- 29444-102
- 29444-100
- 29444-104
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- I-RECs, ISO 14001 - Reynosa