HTS Transwell® 24 Clear Not Treated Reservoir, Sterile, Corning
# de catalogue 29445-096
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Pore size:Open Culture Reservoir
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:29445-096
- Packaging:Individually Wrapped
- Supplier No.:3395
Spécifications
A propos de cet article
For use as reservoir tray with HTS Transwell permeable supports.
- Not treated for cell attachment
- Same footprint as standard 24 well plate but with single rectangular well
Customers Who Bought This Also Bought