Combiplate™ Microstrip® and MultiFrame Plates, 96-Well Format, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
These plates consist of 8- or 12-well strips, either solid or breakable, assembled in a frame
Strips are reliable and feature even coating and washing. A recessed bottom keeps the strip surface clean and scratch free. Strips are available with universal binding or enhanced binding surfaces. Plates and strips are available in clear, black, and or white. Frame is suitable for stacking.
Each frame accommodates a maximum of 96 wells. The reusable frame supports each well individually, allowing for use of the exact number of wells needed. Features include a patented well locking system to keep wells in place, top-quality materials to ensure high optical performance and optimum binding characteristics. Maximum well volume of 400µL.
Customers Who Bought This Also Bought