Corning® Gel-Loading Pipette Tips, Corning
Supplier: Corning
Certificates
About this item
These gel-loading pipette tips feature a capillary end that allows easy access into vertical and horizontal electrophoresis gels.
- Disposable
- Certified RNase-/DNase-free and non pyrogenic
Ordering information: Available in flat and round tip configurations. Flat tips are designed for use in 0.2 and 0.4 mm sequencing gels. Round tips are designed for use in SDS-PAGE 0.5 mm gels.
Customers Who Bought This Also Bought