Mµlti™ MiniFlex™ Pipette Tips, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
Tips feature an elongated, flexible, ultra-thin microcapillary that is ideal for nucleic acid sequencing and protein separation.
- A 2 µl graduation permits easy volume QuikCheck
- Round-ended tip is ideal for SDS-PAGE gels
Flat-ended tip is specially designed for use in 0.4 and 0.2 mm sequencing gels. Tips have a unique universal sealing ring for use with most popular brand research-grade pipettors that deliver volumes from 0.1 to 10 µl.
Certifications: Certified RNase- and DNase-free.
Ordering information: Supplied in polypropylene MµltiRacks of 200 tips.
Customers Who Bought This Also Bought