96-Well Polypropylene PCR Microplates, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
These ultrathin polypropylene PCR plates are available with full, half, or no skirt to facilitate compatibility with a wide range of thermocyclers.
- Polypropylene construction
- Free of RNase, DNase, and human DNA
- Nonpyrogenic
- Available in a variety of skirt options
Plates are RNase-, DNase-, and pyrogen-free, and also free of human DNA.
Ordering information: Half skirt plate 82050-698 is compatible with ABI Sequencing Systems. Half skirt plate 82050-704 is compatible with the ABI PRISM® 7700.
Customers Who Bought This Also Bought